Skip to Content
Merck
All Photos(1)

Documents

21578

Needle Cleaning Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122001

manufacturer/tradename

Hamilton 76620

packaging

1 ea of

General description

Formulated specifically for GC syringes.
Perfect for cleaning small orifices such as FID jets and syringe needles. This kit includes one 70 mL bottle (makes 700 mL of cleaning solution), ten wires in each of five sizes (0.076 to 0.306 mm O.D.), and instructions.

Packaged in a reusable box that prevents wires from being damaged during storage.

Cleaning wires (O.D.)Use to clean needle gauges

0.089 mm26s, 31-33
0.126 mm22s, 25s, 28-30
0.167 mm27
0.207 mm24-26 mm
0.306 mm22, 23 and larger

Application

Needle Cleaning Kit may be used as first aid kit for syringe to insure its accuracy and prolonged life.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analytical chemists at the launch pad.
Bennett, Helein D.
Analytical Chemistry, 49.3, 322A-326A (1977)

Protocols

Syringes are precision instruments and need care. Learn to correctly inspect, use, unclog, handle, clean and store them to ensure good accuracy, reproducibility, and lifetime.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service