19137-U
Hamilton® needles for RN syringes
needle size 22 ga, bevel tip (point style 2), needle L 51 mm (2 in.), for use with 250μL-10mL
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
bevel tip stainless steel needle (point style 2)
needle size
22 ga
feature
bevel tip (point style 2)
packaging
pkg of 6 ea
manufacturer/tradename
Hamilton 7779-01
needle L
51 mm (2 in.)
volume
250-10000 μL
compatibility
for use with 250μL-10mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Hamilton® needles for RN syringes are bevel tip needles, designed for optimum septum penetration and to prevent septum coring.
Legal Information
Hamilton is a registered trademark of Hamilton Co.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service