Skip to Content
Merck
All Photos(1)

Key Documents

16169

Sigma-Aldrich

SODOSIL RM 001

alkaline cleaner

Sign Into View Organizational & Contract Pricing


About This Item

UBA Number:
08050027
UNSPSC Code:
12000000

technique(s)

NMR: suitable

pH

12-14 (20 °C, direct)

density

1.12-1.18 g/mL at 20 °C

Reconstitution

The best cleaning results are achieved with = 2−20% for normal contamination. The cleaning time = 2−24 h depends on the degree of contamination and on the bath temperature. The action of ultra-sonic waves accelerates the cleaning operation considerably as does an increase in temperature. The cleaning solution can be prepared with tap water. The use of demineralisied water results in more optimum utilisation of baths.

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Legal Information

SODOSIL is a trademark of Sigma-Aldrich Chemie GmbH

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

nwg


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service