Skip to Content
Merck
All Photos(1)

Documents

P1246

Sigma-Aldrich

p30

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.54

recombinant

expressed in E. coli

Quality Level

form

solution

storage temp.

−20°C

SMILES string

Cl.CN(C)CCOc1ccc2OCCOc2c1

InChI

1S/C12H17NO3.ClH/c1-13(2)5-6-14-10-3-4-11-12(9-10)16-8-7-15-11;/h3-4,9H,5-8H2,1-2H3;1H

InChI key

VRCSNYKNBODDIM-UHFFFAOYSA-N

Application

p30 is an endogenous truncated form of p39 that associates with Cdk5 to form Nclk, which regulates neurotransmission, axon guidance, central nervous system (CNS) architecture, and pathogenesis. Therefore, p30 is useful in studying neurodegenerative diseases such as Alzheimer′s, Amyotrophic Lateral Sclerosis, and Parkinson′s Disease.

Biochem/physiol Actions

P30 activates GST-Neuronal cyclin-dependent kinase 5 (Cdk5). It also associates with Cdk5 to form an activated neuronal Cdc2-like kinase (Nclk). Product P1246 is recombinant, from bovine and is expressed in E. coli.

Unit Definition

One unit will incorporate 1 picomole of phosphate into histone H1 peptide per minute at pH 7.2 at 30 °C.

Physical form

p30 is supplied in a solution of 25 mM Tris, pH 7.4, 1 mM DTT, and 30% glycerol. (0.5 μg protein/ 25 ul vial)

Preparation Note

Expressed as a GST fusion protein.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

X H Cai et al.
Neuroscience research, 28(4), 355-360 (1997-08-01)
We previously reported that a neuron-specific Cdk5 activator, p35nck5ai, was most prominent in the newborn rat brain. In the adult brain, the expression decreased in most regions except hippocampus and primary olfactory cortex. A novel neuron-specific Cdk5 activator, p39nck5ai, has
Y Honjyo et al.
Neuroreport, 10(16), 3375-3379 (1999-12-22)
We examined the immunohistochemical localization of p39, the regulatory subunit of cyclin-dependent kinase 5 (Cdk5), in the adult rat brain. Cdk5 requires either p39 or p35 to assume its active form in the mammalian CNS. We developed a specific antibody
Shirley B Shelton et al.
Journal of neurochemistry, 88(6), 1313-1326 (2004-03-11)
Cyclin-dependent kinase-5 (CDK5) is predominantly active in the nervous system and it is well established that CDK5 is essential in neuronal development. In addition to its recognized role in development, there is increasing evidence that CDK5 may be involved in
M Zheng et al.
Journal of neurobiology, 35(2), 141-159 (1998-05-15)
The ubiquitously expressed cyclin-dependent kinase 5 (cdk5) is essential for brain development. Bioactivation of cdk5 in the brain requires the presence of one of two related regulatory subunits, p35 and p39. Since either protein alone can activate cdk5, the significance
D Tang et al.
The Journal of biological chemistry, 270(45), 26897-26903 (1995-11-10)
Neuronal Cdc2-like kinase is a heterodimer of Cdk5 and a 25-kDa subunit that is derived from a 35-kDa brain- and neuron-specific protein called the neuronal Cdk5 activator (p35/p25nck5a) (Lew, J., Huang, Q.-Q., Qi, Z., Winkfein, R. J., Aebersold, R., Hunt

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service