Skip to Content
Merck
All Photos(1)

Documents

14482

Supelco

Tris(2-butoxyethyl) phosphate

analytical standard

Synonym(s):

Phosphoric acid tris(2-butoxyethyl) ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)3OCH2CH2O]3P(O)
CAS Number:
Molecular Weight:
398.47
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

13.7 (vs air)

vapor pressure

0.03 mmHg ( 150 °C)

Assay

≥95.0% (GC)

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.435-1.439
n20/D 1.438 (lit.)

bp

215-228 °C/4 mmHg (lit.)

density

1.006 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

SMILES string

CCCCOCCOP(=O)(OCCOCCCC)OCCOCCCC

InChI

1S/C18H39O7P/c1-4-7-10-20-13-16-23-26(19,24-17-14-21-11-8-5-2)25-18-15-22-12-9-6-3/h4-18H2,1-3H3

InChI key

WTLBZVNBAKMVDP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tris(2-butoxyethyl) phosphate belongs to the class of organophosphate triesters, widely used as flame retardants and plasticizers in a variety of commercial products such as textiles, foams, polymers, paints, hydraulic fluids, etc.

Application

Tris(2-butoxyethyl) phosphate may be used as an analytical reference standard for the quantification of the analyte in herring gull eggs, house dust, and urine samples using liquid chromatography technique.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

318.2 °F - closed cup

Flash Point(C)

159 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analysis of organophosphate flame retardant diester metabolites in human urine by liquid chromatography electrospray ionisation tandem mass spectrometry
Eede dVN, et al.
Journal of Chromatography A, 1303, 48-53 (2013)
Da Chen et al.
Journal of chromatography. A, 1220, 169-174 (2011-12-22)
Numerous triester organophosphate flame retardants (OPFRs) have been used for several decades and continue to be used in a variety of commercial products. We developed a sensitive quantitative method for the analysis of, seven non-halogenated, three chlorinated and two brominated
Non-target analysis of household dust and laundry dryer lint using comprehensive two-dimensional liquid chromatography coupled with time-of-flight mass spectrometry
Ouyang X, et al.
Chemosphere, 166(28), 431-437 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service