Skip to Content
Merck
All Photos(1)

Key Documents

07318

Sigma-Aldrich

Magnetic iron oxide nanoparticles

0.8-1.4% solid material basis, 0.5-0.7% Fe basis

Synonym(s):

Iron oxide Nanoparticles, magnetic

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
R$1,111.00
100 ML
R$2,831.00

R$1,111.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 ML
R$1,111.00
100 ML
R$2,831.00

About This Item

UNSPSC Code:
12352302
NACRES:
NA.23

R$1,111.00


Check Cart for Availability

Request a Bulk Order

form

liquid

contains

oleic acid as stabilizer

composition

Fe, 0.5-0.7%
solid material, 0.8-1.4%

concentration

in heptane

diameter

10-40 nm (hydrodynamic diameter, 1% mean volume)

particle size

6.5 nm±3.0 nm (Fe-Oxide-Nucleus)

density

0.69 g/mL at 20 °C

Analysis Note

Z-Average: ~33 nm

related product

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

24.8 °F

Flash Point(C)

-4 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service