Skip to Content
Merck
All Photos(1)

Key Documents

06617

Sigma-Aldrich

Octyl-α-D-glucopyranoside 100 mM solution

Synonym(s):

Octyl-α-D-glucopyranoside solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352201

storage temp.

2-8°C

InChI

1S/C14H28O6/c1-2-3-4-5-6-7-8-19-14-13(18)12(17)11(16)10(9-15)20-14/h10-18H,2-9H2,1H3/t10-,11-,12+,13-,14+/m1/s1

InChI key

HEGSGKPQLMEBJL-RGDJUOJXSA-N


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zahrabatoul Mosapour Kotena et al.
Journal of molecular modeling, 19(2), 589-599 (2012-09-14)
Density functional theory calculations on two glycosides, namely, n-octyl-β-D-glucopyranoside (C(8)O-β-Glc) and n-octyl-β-D-galactopyranoside (C(8)O-β-Gal) were performed for geometry optimization at the B3LYP/6-31G level. Both molecules are stereoisomers (epimers) differing only in the orientation of the hydroxyl group at the C4 position.
Kerry K Karukstis et al.
The journal of physical chemistry. B, 116(12), 3816-3822 (2012-03-06)
When mixed with water, n-octyl β-D-glucoside forms self-assembled nanostructures, several of which are liquid crystalline and all of which depend on the water/glucoside ratio and temperature. For practical use of these phases, a detailed understanding of the conditions under which
Matthew J Ranaghan et al.
The journal of physical chemistry. B, 114(44), 14064-14070 (2010-10-23)
The photochemical and thermal stability of the detergent-solubilized blue- and green-absorbing proteorhodpsins, BPR and GPR, respectively, are investigated to determine the viability of these proteins for photonic device applications. Photochemical stability is studied by using pulsed laser excitation and differential
Jörg Grunenberg
Physical chemistry chemical physics : PCCP, 13(21), 10136-10146 (2011-04-20)
The anomer selectivity of artificial carbohydrate receptors was studied using in silico methods in order to shed light on the thermodynamic driving forces at work during molecular recognition in general. The contributions of relevant intermolecular hydrogen bonds were investigated by
Jun Liu et al.
Biochemistry, 50(24), 5497-5506 (2011-05-17)
The ATP synthase of the alkaliphile Bacillus pseudofirmus OF4 has a tridecameric c-subunit rotor ring. Each c-subunit has an AxAxAxA motif near the center of the inner helix, where neutralophilic bacteria generally have a GxGxGxG motif. Here, we studied the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service