Merck
  • Mutations in a helix-1 motif of the ATP synthase c-subunit of Bacillus pseudofirmus OF4 cause functional deficits and changes in the c-ring stability and mobility on sodium dodecyl sulfate-polyacrylamide gel electrophoresis.
Mutations in a helix-1 motif of the ATP synthase c-subunit of Bacillus pseudofirmus OF4 cause functional deficits and changes in the c-ring stability and mobility on sodium dodecyl sulfate-polyacrylamide gel electrophoresis.
Biochemistry (2011-05-17)
Jun Liu, Oliver J Fackelmayer, David B Hicks, Laura Preiss, Thomas Meier, Eric A Sobie, Terry A Krulwich
ABSTRACT
The ATP synthase of the alkaliphile Bacillus pseudofirmus OF4 has a tridecameric c-subunit rotor ring. Each c-subunit has an AxAxAxA motif near the center of the inner helix, where neutralophilic bacteria generally have a GxGxGxG motif. Here, we studied the impact of four single and six multiple Ala-to-Gly chromosomal mutations in the A16xAxAxA22 motif on the capacity for nonfermentative growth and, for most of the mutants, on ATP synthesis by ADP- and P(i)-loaded membrane vesicles at pH 7.5 and 10.5. Sodium dodecyl sulfate-polyacrylamide gel electrophoresis (SDS-PAGE) analyses of the holo-ATP synthases were used to probe stability of the mutant c-rotors and mobility properties of the c-rotors as well as the monomeric c-subunits that are released from them by trichloroacetic acid treatment. Mutants containing an Ala16-to-Gly mutation exhibited the most severe functional defects. Via SDS-PAGE, most of the mutant c-monomers exhibited increased mobility relative to the wild-type (WT) c-subunit, but among the intact c-rings, only Ala16-to-Gly mutants exhibited significantly increased mobility relative to that of the WT c-ring. The hypothesis that these c-rings have a decreased c-subunit stoichiometry is still untested, but the functional impact of an Ala16-to-Gly mutation clearly depended upon additional Ala-to-Gly mutation(s) and their positions. The A16/20G double mutant exhibited a larger functional deficit than both the A16G and A16/18G mutants. Most of the mutant c-rings showed in vitro instability relative to that of the WT c-ring. However, the functional deficits of mutants did not correlate well with the extent of c-ring stability loss, so this property is unlikely to be a major factor in vivo.
Sigma-Aldrich
Octyl β-D-glucopyranoside, BioXtra, ≥98% (GC)
Sigma-Aldrich
Octyl β-D-glucopyranoside solution, ≥95% (HPLC), 50 % (w/v) in H2O
Sigma-Aldrich
Octyl β-D-glucopyranoside, ≥98% (GC)
Sigma-Aldrich
Octyl-β-D-glucopyranoside 100 mM solution