Skip to Content
Merck
All Photos(1)

Documents

00586

Sigma-Aldrich

Acetone

purum, ≥99.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH3
CAS Number:
Molecular Weight:
58.08
Beilstein:
635680
EC Number:
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:

vapor density

2 (vs air)
2 (vs air)

vapor pressure

184 mmHg ( 20 °C)
184 mmHg

grade

purum

Assay

≥99.0% (GC)

form

liquid

expl. lim.

13.2 %
8 % (lit.)

impurities

≤1% water

refractive index

n20/D 1.359 (lit.)
n20/D 1.359

bp

56 °C/760 mmHg (lit.)
56 °C/760 mmHg (lit.)

mp

-94 °C (lit.)
−94 °C (lit.)

density

0.791 g/mL at 25 °C (lit.)

format

neat

SMILES string

CC(C)=O

InChI

1S/C3H6O/c1-3(2)4/h1-2H3

InChI key

CSCPPACGZOOCGX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Photocatalytic activity of mixtures of TiO2 (P25) with rare earth oxides for the oxidation of acetone has been reported. The vapor-phase Aldol condensation of acetone over MgO promoted with alkali (Li, Na, K and Cs) or alkaline earth (Ca, Sr and Ba) metal ions has been studied. Acetone undergoes Aldol condensation in the presence of catalysts Mg-Al layered double hydroxides (LDH) and Cl- and/or CO32- as compensating anions to afford diacetone alcohol and mesityl oxide. Enantioselective Aldol condensation of acetone with several isatins has been described.

Application

Acetone may be employed in the following studies:
  • As organic solvent to investigate the zeta potential of TiO2 and Al2O3 powders.
  • Synthesis of poly(ortho esters) (POE), biodegradable carriers for drug delivery devices.
  • Spectrophotometric determination of ester groups in lipids.
Acetone′s luminesence intensity is dependent upon the solution components . The absorption of UV light by acetone, results in its photolysis and the production of radials .

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

A Merkli et al.
Journal of biomaterials science. Polymer edition, 4(5), 505-516 (1993-01-01)
Since the late 1970s, three families of poly(ortho esters) (POE) were synthesized to provide bioerodible carriers for drug delivery devices. The most recent POE is a semi-solid polymer with a viscous behavior at room temperature. Polymer synthesis by a transesterification
Location of the shear plane in the electric double layer in an organic medium.
Siffert B, et al.
Journal of Colloid and Interface Science, 163(2), 327-333 (1992)
Base catalysis for the synthesis of α,β-unsaturated ketones from the vapor-phase aldol condensation of acetone.
Di Cosimo JI, et al.
Applied Catalysis A: General, 137(1), 149-166 (1996)
A simplified spectrophotometric determination of ester groups in lipids.
F SNYDER et al.
Biochimica et biophysica acta, 34, 244-245 (1959-07-01)
Gianluigi Luppi et al.
The Journal of organic chemistry, 70(18), 7418-7421 (2005-08-27)
[reaction: see text] The aldol condensation of acetone with several isatins is described. The desired compound was obtained in quantitative yield and with good enantioselectivities up to 77%. The best results were obtained with 10 mol % H-D-Pro-L-beta3-hPhg-OBn as a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service