EX0290
Ethyl alcohol, pure
94.9-96.0% (GC), suitable for HPLC, 190 Proof, Meets ACS Specifications
Synonym(s):
Ethanol, Ethyl alcohol
About This Item
Recommended Products
product name
Ethyl Alcohol, Pure, 190 Proof, HPLC, Meets ACS Specifications
vapor density
1.59 (vs air)
Quality Level
vapor pressure
44.6 mmHg ( 20 °C)
Assay
94.9-96.0% (GC)
form
liquid
autoignition temp.
683 °F
expl. lim.
19 %, 60 °F
technique(s)
HPLC: suitable
impurities
≤0.0002 meq/g Titrable base
≤0.0005 meq/g Titrable acid
≤0.1% Methanol content (CH3OH)
evapn. residue
≤0.001%
color
APHA: ≤10
refractive index
n20/D 1.3600 (lit.)
pH
7.0 (20 °C, 10 g/L in H2O)
bp
78 °C (lit.)
mp
−114 °C (lit.)
transition temp
flash point 17 °C
density
0.789 g/mL at 25 °C (lit.)
UV absorption
λ: 210 nm Amax: ≤0.4
λ: 220 nm Amax: ≤0.25
λ: 230 nm Amax: ≤0.15
λ: 240 nm Amax: ≤0.05
λ: 270-400 nm Amax: ≤0.01
shipped in
ambient
storage temp.
room temp
SMILES string
CCO
InChI
1S/C2H6O/c1-2-3/h3H,2H2,1H3
InChI key
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Analysis Note
Color (APHA): 10APHA max
Evaporation residue: 0.001% max
Form: Clear liquid
Methanol content (CH3OH): 0.1% max
Solubility in water: Passes test
Substances darkened by sulfuric acid: Passes test
Substances reducing permanganate: Passes test
Titrable acid: 0.0005meq/g max
Titrable base: 0.0002meg/g max
UV Abs. at 210 nm: 0.4AU max
UV Abs. at 220 nm: 0.25AU max
UV Abs. at 230 nm: 0.15AU max
UV Abs. at 240 nm: 0.05AU max
UV Abs. at 270 - 400 nm: 0.01AU max
Footnote
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
62.6 °F
Flash Point(C)
17 °C
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service