Skip to Content
Merck
All Photos(1)

Key Documents

24-1400

Sigma-Aldrich

Phenol

SAJ first grade, ≥98.0%

Synonym(s):

Hydroxybenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5OH
CAS Number:
Molecular Weight:
94.11
Beilstein:
969616
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
grade:
SAJ first grade
bp:
182 °C (lit.)
vapor pressure:
0.09 psi ( 55 °C)
0.36 mmHg ( 20 °C)
Pricing and availability is not currently available.

grade

SAJ first grade

vapor density

3.24 (vs air)

vapor pressure

0.09 psi ( 55 °C)
0.36 mmHg ( 20 °C)

Assay

≥98.0%

form

solid

autoignition temp.

1319 °F

expl. lim.

8.6 %

availability

available only in Japan

dilution

(for analytical testing)

pH

~5 (20 °C, 50 g/L)

bp

182 °C (lit.)

mp

40-42 °C (lit.)

density

1.071 g/mL at 25 °C (lit.)

SMILES string

Oc1ccccc1

InChI

1S/C6H6O/c7-6-4-2-1-3-5-6/h1-5,7H

InChI key

ISWSIDIOOBJBQZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Muta. 2 - Skin Corr. 1B - STOT RE 2

Target Organs

Nervous system,Kidney,Liver,Skin

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

177.8 °F - closed cup

Flash Point(C)

81 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yanuardi Raharjo et al.
Materials science & engineering. C, Materials for biological applications, 103, 109722-109722 (2019-07-28)
In this work, the novel imprinted zeolite (IZ) was synthesized, and its properties and performance in terms of adsorption of p-Cresol, which represent the protein-bounded uremic toxins in aqueous phase under phosphate buffer saline, were studied and compared with the
Octavian Bucur et al.
Scientific reports, 5, 9893-9893 (2015-05-12)
Recombinant soluble TRAIL and agonistic antibodies against TRAIL receptors (DR4 and DR5) are currently being created for clinical cancer therapy, due to their selective killing of cancer cells and high safety characteristics. However, resistance to TRAIL and other targeted therapies
Christos Andreadis et al.
Nucleic acids research, 42(20), 12650-12667 (2014-10-11)
DNA damage response and repair proteins are centrally involved in genome maintenance pathways. Yet, little is known about their functional role under non-DNA damage-inducing conditions. Here we show that Rad9 checkpoint protein, known to mediate the damage signal from upstream
Juston C Weems et al.
The Journal of biological chemistry, 290(24), 15030-15041 (2015-04-17)
Elongin A performs dual functions in cells as a component of RNA polymerase II (Pol II) transcription elongation factor Elongin and as the substrate recognition subunit of a Cullin-RING E3 ubiquitin ligase that has been shown to target Pol II
Mayilvahanan Shanmugam et al.
PloS one, 10(2), e0117487-e0117487 (2015-02-24)
Aggregatibacter actinomycetemcomitans a causative agent of periodontal disease in humans, forms biofilm on biotic and abiotic surfaces. A. actinomycetemcomitans biofilm is heterogeneous in nature and is composed of proteins, extracellular DNA and exopolysaccharide. To explore the role played by the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service