Skip to Content
Merck
All Photos(1)

Key Documents

279382

Sigma-Aldrich

Ethyl acetate-1-13C

99 atom % 13C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH313CO2C2H5
CAS Number:
Molecular Weight:
89.10
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

vapor density

3 (vs air)

Quality Level

vapor pressure

73 mmHg ( 20 °C)

isotopic purity

99 atom % 13C

Assay

99% (CP)

expl. lim.

11.4 %

refractive index

n20/D 1.372 (lit.)

bp

76-77 °C (lit.)

mp

-84 °C (lit.)

density

0.912 g/mL at 25 °C

mass shift

M+1

SMILES string

CCO[13C](C)=O

InChI

1S/C4H8O2/c1-3-6-4(2)5/h3H2,1-2H3/i4+1

InChI key

XEKOWRVHYACXOJ-AZXPZELESA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

26.6 °F - closed cup

Flash Point(C)

-3 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service