Skip to Content
Merck
All Photos(1)

Key Documents

19-1010

Sigma-Aldrich

Manganese standard solution

suitable for atomic absorption spectrometry, 1000 ppm Mn+2, 1 mg/mL Mn+2

Synonym(s):

Manganese(II) chloride solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MnCl2
CAS Number:
Molecular Weight:
125.84
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
grade:
SAJ special grade
form:
liquid
Pricing and availability is not currently available.

grade

SAJ special grade

form

liquid

availability

available only in Japan

concentration

1 mg/mL Mn+2
1000 ppm Mn+2

suitability

suitable for atomic absorption spectrometry

SMILES string

Cl[Mn]Cl

InChI

1S/2ClH.Mn/h2*1H;/q;;+2/p-2

InChI key

GLFNIEUTAYBVOC-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jerome A Roth et al.
Neurotoxicology, 35, 121-128 (2013-01-15)
Chronic exposure to Mn results in the development of a neurological disorder known as manganism characterized by neurological deficits resembling that seen in Parkinsonism. Although dopaminergic neurons within the nigrostriatal pathway appear intact, Mn-induced irregularities in DA transmission have been
Anne Bertrand et al.
NeuroImage, 64, 693-702 (2012-09-11)
The impairment of axonal transport by overexpression or hyperphosphorylation of tau is well documented for in vitro conditions; however, only a few studies on this phenomenon have been conducted in vivo, using invasive procedures, and with contradictory results. Here we
A G Kanthasamy et al.
Toxicology letters, 214(3), 288-295 (2012-09-22)
The role of normal cellular prion protein (PrP) remains to be fully elucidated; however, the protein is crucial for the infection and progression of prion diseases. Recent evidence indicates that PrP is a metalloprotein since the octapeptide repeat sequences in
Barry J Bowman et al.
Eukaryotic cell, 11(11), 1362-1370 (2012-09-18)
The pmr gene is predicted to encode a Ca(2+)-ATPase in the secretory pathway. We examined two strains of Neurospora crassa that lacked PMR: the Δpmr strain, in which pmr was completely deleted, and pmr(RIP), in which the gene was extensively
M I Mokgobu et al.
Inhalation toxicology, 24(10), 634-644 (2012-08-22)
Although pro-inflammatory mechanisms have been implicated in the pathogenesis of manganese (Mn²⁺)-related neurological and respiratory disorders, relatively little is known about the potential of this metal to interact pro-oxidatively with human phagocytes. The primary objective of the current study was

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service