Merck
  • Manganese promotes increased formation of hydrogen peroxide by activated human macrophages and neutrophils in vitro.
Manganese promotes increased formation of hydrogen peroxide by activated human macrophages and neutrophils in vitro.
Inhalation toxicology (2012-08-22)
M I Mokgobu, R Anderson, H C Steel, M C Cholo, G R Tintinger, A J Theron
ABSTRACT
Although pro-inflammatory mechanisms have been implicated in the pathogenesis of manganese (Mn²⁺)-related neurological and respiratory disorders, relatively little is known about the potential of this metal to interact pro-oxidatively with human phagocytes. The primary objective of the current study was to investigate the effects of Mn²⁺ as MnCl₂ (0.5-100 µM) on the generation of the reactive oxygen species (ROS), superoxide, hydrogen peroxide (H₂O₂), and hypohalous acids by isolated human blood neutrophils and monocyte-derived macrophages following activation of these cells with the chemotactic tripeptide, FMLP (1 µM), or the phorbol ester, PMA (25 ng/mL). Generation of ROS was measured using the combination of oxygen consumption, lucigenin/luminol-enhanced chemiluminescence, spectrofluorimetric detection of oxidation of 2,7-dichlorodihydrofluorescein, radiometric assessment of myeloperoxidase (MPO)-mediated protein iodination, release of MPO by ELISA, and spectrophotometric measurement of nitrite formation. Treatment of activated neutrophils with either FMLP or PMA resulted in significantly decreased reactivity of superoxide in the setting of increased formation of H₂O₂ and MPO-mediated iodination, with no detectable effects on either oxygen consumption or MPO release. Similar effects of the metal with respect to superoxide reactivity and H₂O₂ formation were observed with activated macrophages, while generation of NO was unaffected. Taken together with the findings of experiments using cell-free ROS-generating systems, these observations are compatible with a mechanism whereby Mn²⁺, by acting as a superoxide dismutase mimetic, increases the formation of H₂O₂ by activated phagocytes. If operative in vivo, this mechanism may contribute to the toxicity of Mn²⁺.
Sigma-Aldrich
Manganese(II) chloride tetrahydrate, BioUltra, for molecular biology, ≥99.0% (KT)
Sigma-Aldrich
Manganese(II) chloride, beads, 98%
Sigma-Aldrich
Manganese(II) chloride solution, BioReagent, for molecular biology, storage temp.:room temp
Sigma-Aldrich
Manganese(II) chloride, powder and chunks, ≥99% trace metals basis
Sigma-Aldrich
Manganese(II) chloride tetrahydrate, 99.99% trace metals basis
Sigma-Aldrich
Manganese(II) chloride tetrahydrate, ReagentPlus®, ≥99%
Sigma-Aldrich
Manganese(II) chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Manganese(II) chloride tetrahydrate, meets USP testing specifications
Sigma-Aldrich
Manganese(II) chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Manganese(II) chloride tetrahydrate, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Manganese(II) chloride tetrahydrate, ACS reagent, ≥98%
Sigma-Aldrich
Manganese(II) chloride 0.1 M solution