Skip to Content
Merck
All Photos(8)

Key Documents

ZIX7010P0C

Milli-Q

Milli-Q® IX Pure Water System with E-POD® Dispenser

greener alternative

input: potable tap water, output: type 2 water (> 5 MΩ·cm), The most advanced pure water system for the production of Elix® quality water at a flow rate of 10 L/h, with E-POD® pure water dispenser.

Synonym(s):

Milli-Q® IX 7010 Pure Water System with E-POD® Dispenser

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104202
NACRES:
JA.13

Finding the right water system for your lab? Use our selection guide to determine the kind of system that would fit your needs at the lab or contact a specialist for further assistance.

AC/DC input

100 V / 240 V, 50-60 Hz

Quality Level

product line

Milli-Q® IX

packaging

pkg of 1 unit (System + E-POD®)

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

parameter

10 L/hr flow rate
200 L/day max. usage
29.7 kg operating weight (65 lb)
~2 L/min distribution flow rate

H × D × W

70.2 cm (27.6 in.) × 27 cm (10.6 in.) × 21.1 cm (8.3 in.), E-POD®

system H × W × D

49.8 cm (19.6 in.) × 37.5 cm (14.8 in.) × 38 cm (15 in.)

input

feed water nature potable tap water

output

product water quality: type 2 water (> 5 MΩ·cm)

conductivity

0.2 μS/cm at 25 °C (water output)

greener alternative category

Looking for similar products? Visit Product Comparison Guide

General description

This item number includes the System and one E-POD® dispenser.

Milli-Q® IX 7010 Pure Water System
The Milli-Q® IX 7010 system is a compact, ergonomic, mercury-free and intelligent water purification solution that delivers consistently high quality pure water on demand.

The system features three separate and distinct components:
  • The pure water production unit offers flexible installation and simplified maintenance.
  • The intelligent pure water storage tank allows maximum protection from any external source of contamination.
  • The redesigned E-POD® point-of-delivery unit provides convenient, rapid access to pressurized pure water. A large touch screen allows for intuitive interaction and displays essential data for complete user reassurance. Up to two E-POD® dispensers can be used with each IX production unit at different locations within the same laboratory, or in adjacent laboratories.


E-POD® Dispenser
Always have rapid and convenient access to pressurized Type 2 pure water. This ergonomic dispenser is compact and intuitive to use, supporting an efficient working environment. Essential system functions remain close at hand on the E-POD® touchscreen interface.

Application

The Milli-Q® IX 7010 Water Purification System produces pure (Type 2) water.

  • For critical lab applications such as: microbiology culture media preparation, staining solutions for histology and cytology, immunohistochemistry (IHC), electrophoresis gel and buffers, western blotting, immunoassays (ELISA, RIA), drug dissolution testing, biological oxygen demand (BOD), chemical oxygen demand (COD), UV/Vis spectroscopy, titration, etc. It can also be used to prepare samples for laser fluorometry, in the synthesis and characterization of membranes for fuel cells,[1] or in the synthesis of nanocomposites by electrochemistry.[2]
  • For general lab applications: Sample preparation (dilution, extraction...), buffer and reagent preparation, glassware rinsing, etc.
  • To feed laboratory instruments like autoclaves, dishwashers, weathering and stability test chambers, clinical analyzers and slide stainers, hydrogen generators, and ultrapure water systems[3] (e.g. Milli-Q® IQ 7000 system).


E-POD® Dispenser
  • The E-POD® remote dispenser is designed to provide dispensing Type 2 pure water flexibility, with variable flow rates and volumetric dispensing.

Features and Benefits

Milli-Q® IX 7010 Pure Water System
  • Delivers pure water quality that meets Pharmacopeia and ISO requirements.
  • Easy to use with carefree maintenance.
  • Delivers reliable Type 2 pure water with continuous quality monitoring.
  • Compact with space-saving installation options.
  • Mercury-free ECH2O® lamps use UVC LED technology to inactivate bacteria.
  • Uses IPAK Gard® pre-treatment cartridge protects the reverse osmosis (RO) membrane to improve system performance.
  • Elix® electrodeionization (EDI) module delivers consistently superior quality pure water with no maintenance and at low running costs.
  • Intelligent pure water storage solution provides multi-targeted protection from bacterial, particulate, and CO2 contamination by using bactericidal UV lamp, a vent filter, automatic sanitization module (ASM), and an overflow sensor.
  • Automatic e-record archiving.
  • Integrated, simplified data management.

E-POD® Dispenser
  • Provides pressurized pure water, up to 2 L/min.
  • 3 different final filters can be connected to target specific contaminants and remove them right when dispensing pure water.
  • 5" touchscreen with bright, colored LCD display is easy to read and convenient to use, even with gloves on. It is located at eye level and orientable (120°).
  • Navigation in operating and dispensing modes is intuitive, without spending time going through multiples menus to find the information.
  • Graphic icons on the display allow all users to quickly and easily operate the system.
  • Comprehensive information displayed on the screen (water quality, system status, tank level, operating mode, volume dispensed) allows to always dispense the expected water quality.
  • Two modes to dispense pure water:
  • Variable flow (2 different flows, thanks to the dispensing wheel: low flow and full flow up to 2 L/min).
  • Volumetric water dispensing automatically delivers a specific water volume, from 20 mL to 100 L (in 1 mL increments).
  • The water dispenser has more than 1,500 positions (up to 30 horizontal notched positions, 360° around, and 54 vertical notched positions). Hence, daily operations are easier because the dispenser is well adapted to the size and shape of all the containers used in the laboratory.
  • Can be installed on a wall-mounted horizontal articulated extension, allowing for maximum bench space.

MyMilli-Q Digital Services
Be fully supported by MyMilli-Q Remote Care online monitoring and service capability, for your lab′s maximum productivity.
  • Immediate and customizable notifications of system alerts and alarms 24/7, by e-mail and SMS.
  • Real-time system information, performance, water quality data and more on your computer or mobile device.
  • Rapid, online diagnostics – and even remote repairs possible – by our service team.
  • Full data traceability and effortless data access and retrieval capabilities facilitate audit preparation and lab accreditation.
  • Online management of your service and consumable contracts.
A Greener Alternative Product: The Milli-Q® IX 7010 system has been evaluated on 7 dimensions that have a critical impact on Global Warming: Energy & Emissions, Water, Packaging, Materials, Suppliers & Manufacturing, Circular Economy, and Usability & Innovation. Officially awarded the Greener Alternative Product label, we certify that the Milli-Q® IX 7010 system uses less electricity, water and plastic, and is completely mercury free. View the Design for Sustainaibility (DfS) scorecard for the Milli-Q® IX 7010 system.

Other Notes

Components required but not included:
  • A specific country box with the user manual in local language and a power cord adapted to the local electrical network needs to be ordered in addition to this catalog number.
  • The indicated price currently corresponds to the above catalog number only.
  • It is subject to change at any time without notice.
  • This catalog number does not include consumables, accessories and other products and/or services that may be required for the installation and the correct operation of the water purification unit over time. A Milli-Q® IQ/IX storage tank 25 L, 50 L or 100 L and any additional E-POD® remote dispenser needs to be ordered in addition to this catalog number.
  • Because our customers each have specific purified water needs, we recommend that you have your purchase validated by one of our specialists. We remain at your disposal to advise you, do not hesitate to contact us!


For more information on this product please contact our Technical Service Team

Legal Information

E-POD is a registered trademark of Merck KGaA, Darmstadt, Germany
ECH2O is a registered trademark of Merck KGaA, Darmstadt, Germany
ELIX is a registered trademark of Merck KGaA, Darmstadt, Germany
IPAK GARD is a registered trademark of Merck KGaA, Darmstadt, Germany
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
MyMilli-Q is a trademark of Merck KGaA, Darmstadt, Germany

accessory

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guanidinium based blend anion exchange membranes for direct methanol alkaline fuel cells (DMAFCs)
Sajjad S D, et al.
Journal of Power Sources, 300, 95-103 (2015)
Corrosion inhibiting action of Ni?Mo alloy coatings in the presence of mixed metal oxide nanocomposites
Goveas J J, et al.
New. J. Chem., 42, 13660-13666 (2018)
Takahiro Yamaguchi et al.
Journal of agricultural and food chemistry, 63(21), 5133-5140 (2015-02-07)
This study involved the development of a multiresidue method for the rapid analysis of 43 antibiotics in meats using ultrahigh-performance liquid chromatography-tandem mass spectrometry. This method was performed using dispersive-solid phase extraction, which is able to analyze 20 samples within

Articles

Reliable histology staining requires consistent quality pure water. See examples of GMS and H&E stains performed with various water contaminants and comparing tap vs Type 2 water.

Elemental impurities in pharmaceutical drugs present a risk to human health and are regulated. Milli-Q® ultrapure water purification systems comply with the requirements for trace element analyses.

Learn how water impurities can affect pH. Water quality defined as pure water or Type 2 water is recommended to prepare buffer solutions or measure pH.

Compare RO vs. EDI vs. ion exchange resins to remove silica from water for silica-sensitive applications (e.g. microelectronics, testing chambers, autoclaves, sterilizers, …).

See All

Related Content

Discover Milli-Q® IX systems for high-purity water. Ideal for general lab & sensitive techniques. Reliable Efficient Innovative. A Greener Alternative.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service