Skip to Content
Merck
All Photos(1)

Key Documents

MERS03PED

Millipore

Foot Pedal Accessory for Millicell® ERS 3.0

Optional external foot pedal or switch for Millicell ERS 3.0 Voltohmmeter.

Synonym(s):

Foot Pedal Accessory for Millicell® ERS 3.0, Foot pedal, foot switch, hands-free switch

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000
NACRES:
NB.24

product line

Millicell®

General description

The Millicell® ERS 3.0 Digital VoltOhmMeter reliably measures membrane potential and resistance of epithelial and endothelial cells in culture. This device qualitatively measures cell monolayer health and cell confluence. The foot switch/pedal accessory can be attached to the Millicell® ERS 3.0 system to achieve hands-free operation.

Application

To study endothelial/epithelial barrier models (like the blood-brain barrier models, gastrointestinal tract models, and pulmonary models), To study efficacy of drugs, To evaluate pharmacological toxicities

Features and Benefits

  • Allows hands-free operation of Millicell® ERS 3.0 Digital VoltOhmMeter
  • Reduces chances of cell contamination due to frequent handling of cell culture dishes

Legal Information

Millicell is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Find answers to frequently asked questions about the Millicell® ERS 3.0, from probe depth adjustment to instrument maintenance and troubleshooting.

Related Content

Intuitive lab instruments, ergonomic tools, accessible data, and forward-thinking solutions for research, biotech, and pharma laboratories.

Streamline TEER data capture and analysis with user-friendly enhancements including an intuitive touchscreen interface, standing in-well probe, and automatic data logging.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service