Skip to Content
Merck
All Photos(6)

Documents

PLRP24H48

Millipore

Millicell® 24 well hanging cell culture inserts

pore size 1.0 μm, translucent PET membrane, diam. 6.5 mm, Tissue Culture (TC)-treated surface, sterile

Synonym(s):

permeable inserts, tissue culture inserts, translucent membrane insert, high pore density insert

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122107
NACRES:
NB.13

material

polystyrene housing
translucent PET membrane

Quality Level

sterility

γ-irradiated
sterile

feature

22x106 pores/cm2

packaging

pack of 48 ea (individually blister packaged)

manufacturer/tradename

Millicell®

parameter

max. temp. at 37 °C

technique(s)

drug transporter assay: suitable
permeability assay: suitable

H

15.5 mm

diam.

6.5 mm

insert maximum volume

0.4 mL

insert working volume

0.2 mL

membrane thickness

9-16 μm

size

24 wells

surface area

0.3 cm2

color

translucent membrane

pore size

1.0 μm pore size

binding type

Tissue Culture (TC)-treated surface

General description

Millicell® cell culture inserts with polyethylene terephthalate (PET) membrane are ideal for the growth and differentiation of various cell types.

This individually sterile-packaged insert has an high pore density, translucent 1.0 μm pore size membrane and fits in most 24-well tissue culture plates.

Application

  • Barrier Assays
  • Cell Proliferation
  • Cell Surface Receptors
  • Co-Culture
  • Epithelial Cell Growth
  • Feeder Layers
  • Fluorescent Detection / Immunohistochemistry
  • In Vitro Fertilization
  • In Vitro Toxicology
  • Microbial Attachment
  • Polarized Protein Secretions
  • Polarized Uptake
  • Transport/Permeability

Features and Benefits

  • Optically translucent TC-treated PET membrane
  • High pore density (22 million pores/cm2)
  • Compatible with most 24-well plates
  • Apical assist rim to prevent membrane puncture or disruption of cells
  • Easily access the basolateral side between the flanges

Legal Information

Millicell is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

How to set up barrier formation and permeability assays using cell culture inserts and TEER measurements.

Discover tips for choosing the best plate to use with Millicell® hanging inserts for your applications, including barrier formation and cell imaging.

Related Content

Streamline TEER data capture and analysis with user-friendly enhancements including an intuitive touchscreen interface, standing in-well probe, and automatic data logging.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service