MABX8332
Anti-pan Plasmodium LDH Antibody, (1mg)
clone 2, Chemicon®, from mouse
Synonym(s):
pan Plasmodium LDH
About This Item
Recommended Products
biological source
mouse
antibody form
purified antibody
antibody product type
primary antibodies
clone
2, monoclonal
species reactivity
Plasmodium falciparum, Plasmodium ovale, Plasmodium malariae, parasites, Plasmodium vivax
manufacturer/tradename
Chemicon®
technique(s)
ELISA: suitable
immunodiffusion: suitable
isotype
IgG1
General description
Specificity
Immunogen
Application
Infectious Diseases
Quality
Physical form
Storage and Stability
Other Notes
Legal Information
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service