Skip to Content
Merck
All Photos(1)

Key Documents

MABX8322

Sigma-Aldrich

Anti-Plasmodium vivax LDH Antibody, (1mg)

clone 2, Chemicon®, from mouse

Synonym(s):

Plasmodium vivax LDH

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

antibody form

purified antibody

antibody product type

primary antibodies

clone

2, monoclonal

purified by

affinity chromatography

species reactivity

parasites, Plasmodium vivax

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunodiffusion: suitable

isotype

IgG1

General description

Malaria is a mosquito-borne infectious disease caused by parasitic microorganisms in the Plasmodium group. Plasmodium vivax (Pv) still causes serious disease but not as acute as Plasmodium falciparum (Pf). Pv LDH is the Pv-specific plasmodium lactate dehydrogenase, an intracellular metabolic enzyme.

Specificity

Plasmodium vivax LDH

Immunogen

Recombinant P. vivax LDH

Application

Detect malaria using mouse monoclonal Anti-Plasmodium vivax LDH, Cat No MABX8322-1KC, validated for use in ELISA and Lateral Flow Assay.
Research Category
Infectious Diseases
Suggested antibody pairs MABX8321-1KC (labeling) and MABX8322-1KC (coating)

Quality

Detect malaria using mouse monoclonal Anti-Plasmodium vivax LDH, Cat No MABX8322-1KC, validated for use in ELIS

Physical form

Protein G purification
Purified mouse monoclonal antibody in buffer containing 50 mM NaCL, 10 mM PBS, pH 8.0, 0.1% sodium azide (NaN3)

Storage and Stability

2-8C. For long-term storage, freeze at -20C.

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service