Skip to Content
Merck
All Photos(3)

Documents

MABC631

Sigma-Aldrich

Anti-KLF4 Antibody, clone 1E6

ascites fluid, clone 1E06, from mouse

Synonym(s):

Krueppel-like factor 4, Epithelial zinc finger protein EZF, Gut-enriched krueppel-like factor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

1E06, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... KLF4(9314)

General description

Krueppel-like factor 4 (KLF4), which is also known as epithelial zinc finger protein (EZF) and gut-enriched krueppel-like factor (GKLF), is a developmentally regulated transcription factor that plays an important role in maintaining the pluripotent state in embryonic stem cells (ESCs) and is used as a marker of the pluripotent state for cells in culture. It is required for the differentiation of epithelial cells and the establishment of the barrier function of the skin, and KLF4 is also expressed during skeletal and kidney development. KLF4 is highly expressed in the skin, lung, and intestine along the epithelial border. This nuclear localized protein can act as both a tumor suppressor and activator depending upon its environment, but its most prominent function is growth suppression. Interestingly, increased expression of KLF4 leads to an increase in expression of P21WAF1/CIP1 which is an important checkpoint protein that inhibits cell cycle progression and is essential in mediating the cell cycle arrest at both the G1/S and G2/M boundaries. Additionally, KLF4 mediates the normal functions of the tumor suppressor p53 as well. Mutations thus in KLF4 lead to unchecked growth and tumor formation, and because of its expression and involvement within the gut, KLF4 is often mutated in many colon cancer cell lines and the lack of KLF4 expression is correlated with colorectal cancer pathogenesis. Elevated expression of KLF4 gene expression has also been associated with gastric cancer, breast cancer, and squamous cell carcinomas. EMD-Millipore’s Anti-KLF4 mouse monoclonal antibody, clone 1E6, has been tested in western blot against recombinant hKLF4 expressed cell lysates and in immunocytochemistry on paraffin embedded human lung and colon cancer tissue. The clone has also been tested in fluorescent immunocytochemistry on the human esophageal squamous carcinoma cell line Eca109 in culture.

Immunogen

Purified recombinant fragment of human KLF4 expressed in E. coli.

Application

Detect KLF4 using this mouse monoclonal antibody, Anti-KLF4 Antibody, clone 1E6 validated for use in IHC & western blotting.
Immunofluorescence Analysis: A 1:200-1,000 dilution from a representative lot detected KLF4 in ECA109 cells.

Western Blotting Analysis: A 1:3,000 dilution from a representative lot detected KLF4 in transfected HEK293 cell lysates.

Optimal working dilutions must be determined by end user.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional

Quality

Evaluated by Immunohistochemistry in human lung cancer and colon tissues.

Immunohistochemistry Analysis: A 1:200-1,000 dilution from a representative lot detected KLF4 in human lung cancer and colon tissues.

Optimal working dilutions must be determined by end user.

Target description

~55 kDa observed. Uncharacterized bands may appear in some lysate(s). Uniprot describes 3 isoforms between ~46 kDa and ~55 kDa

Physical form

Mouse monoclonal IgG1 ascitic fluid containing up to 0.1% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
Human lung cancer and colon tissues

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service