Recommended Products
form
powder
color
off-white
shipped in
ambient
storage temp.
room temp
General description
Celite® 545 is a flux calcined diatomaceous earth with medium permeability applicable as filter aid for rapid filtration in aqueous solution. It is also suitable for highly viscous liquids such as casein, pectin, adhesives, greases, waxes and tars, syrups and synthetic resins.
Application
Celite® 545 can be used as a clarifying agent to break or mitigate colloids in the turbid solution. It is also used as a short filtration pad to remove unreacted inorganic salts present in the reaction mixture before the chromatographic separation of dissolved compounds.
Analysis Note
Color: Off-white
Form: Powder
Form: Powder
Legal Information
Celite is a registered trademark of Imerys Minerals California, Inc.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
STOT RE 1 Inhalation
Target Organs
Lungs
Storage Class Code
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Industrial clarification of wheat-based distillers? solubles and thin stillage
Industrial Crops and Products, 109, 828-835 (2017)
Preparation of phenols by phthaloyl peroxide-mediated oxidation of arenes
Nature Protocols, 9(11), 2624-2629 (2014)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service