Skip to Content
Merck
All Photos(1)

Documents

22142

Sigma-Aldrich

Celite® 577 fine

filter aid, calcined

Synonym(s):

Diatomaceous earth, calcined

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
11101527
PubChem Substance ID:
NACRES:
SB.54

form

powder

quality

calcined

composition

Al2O3, 4.0%
CaO, 0.6%
Fe2O3, 1.1%
Na2O + K2O, 1.2%
SiO2, 91.5%

loss

≤2.0% loss on ignition, 900 °C

particle size

≤40.0 μm (81% (acc. to Cilas))

pH

<7.0 (25 °C, 10% in aq. suspension)

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Caution

Not Intended for cGMP Parenteral Pharmaceutical Manufacturing

Legal Information

Celite is a registered trademark of Imerys Minerals California, Inc.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anhydrides as acylating agents in lipase-catalyzed stereoselective esterification of racemic alcohols.
Bianchi D, et al.
The Journal of Organic Chemistry, 53(23), 5531-5534 (1988)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service