B311-150-S
Tryptic Soy Broth Ready-to-use Bottles
bottle capacity 300 mL, bottle filling volume 150 mL, closure type, crimp cap with septum, box of 12 bottles
Synonym(s):
CASO Broth, Casein Soy Broth, Casein Soyabean Digest Broth, TSB, Tryptone Soy Broth
About This Item
Recommended Products
Agency
EP 2.6.13 (
)
JP 4.06
USP 71
Quality Level
sterility
sterile; autoclaved
form
liquid
feature
closure type crimp cap with septum
packaging
box of 12 bottles
technique(s)
microbiological culture: suitable
sterility testing: suitable
bottle capacity
300 mL
bottle filling volume
150 mL
pH
7.3±0.2 (25 °C)
application(s)
food and beverages
pharmaceutical
storage temp.
2-25°C
suitability
nonselective for
Looking for similar products? Visit Product Comparison Guide
General description
Application
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Resp. Sens. 1
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service