Skip to Content
Merck
All Photos(1)

Key Documents

Z699209

Trypticase Soy Broth (TSB) Double strength, Bottled Broth

Synonym(s):

agar and broth, media broth, trypticase soy

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
R$330.00

R$330.00


Check Cart for Availability


Select a Size

Change View
1 EA
R$330.00

About This Item

UNSPSC Code:
41111700
NACRES:
NB.79

R$330.00


Check Cart for Availability

packaging

bottle of 1 × 100 mL

manufacturer/tradename

Maine Mfg 10496708

application(s)

food and beverages

storage temp.

2-8°C

General description

TSB is a medium that will support the growth of a wide variety of microorganisms including aerobic, facultative, and anaerobic bacteria and fungi.

Other Notes

Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service