Skip to Content
Merck
All Photos(2)

Documents

AB16969

Sigma-Aldrich

Anti-Caspase 9 Antibody, proform & lg. subunit

Chemicon®, from rabbit

Synonym(s):

ICE-LAP6, Mch-6, Apaf-3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human

should not react with

rat

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CASP9(842)

Specificity

Apoptosis is related to many diseases and induced by a family of cell death receptors and their ligands. Cell death signals are transduced by death domain containing adapter molecules and members of the caspase family of proteases. A novel member in the caspase family was recently identified and designated ICE-LAP6, Mch6, and Apaf-3 (Duan et al. 1996; Srinivasula et al. 1996; Nijhawan et al. 1997). Caspase-9 and Apaf-1 bind to each other, which leads to caspase-9 activation (Nijhawan et al. 1997). Caspase-9 is also activated by granzyme B and CPP32 (Duan et al. 1996; Srinivasula et al. 1996). Activated caspase-9 cleaves and activates caspase-3 that is one of the key proteases, being responsible for the proteolytic cleavage of many key proteins in apoptosis (Nijhawan et al. 1997). Caspase-9 plays a central role in cell death induced by a wide variety of apoptosis inducers including TNF-alpha, TRAIL, anti-CD-95, FADD, and TRADD (Pan et al. 1998). Caspase-9 is expressed in a variety of human tissues (Duan et al. 1996; Srinivasula et al. 1996).

Immunogen

Epitope: proform & lg. subunit
Synthetic peptide corresponding to amino acids 41-56 of human caspase-9 (Duan et al. 1996; Srinivasula et al. 1996; Nijhawan et al. 1997). Recognizes proform (46 kDa); sequence analysis predicts cross-reactivity with mouse caspase-9 (>90%).

Application

Detect Caspase 9 using this Anti-Caspase 9 Antibody, proform & lg. subunit validated for use in WB.
Research Category
Apoptosis & Cancer

Metabolism
Research Sub Category
Caspases

Enzymes & Biochemistry
Western blot: 1:1,000 to 1:2,000

Whole cell lysate from HeLa cells can be used as a positive control and a 46 kDa band should be detected.

Optimal working dilutions must be determined by end user.

Linkage

Replaces: 04-443

Physical form

Format: Purified
Immunoaffinity chromatography purified immunoglobulin. Liquid in PBS, containing 0.02% sodium azide.

Storage and Stability

The undiluted antibody solution is stable for approximately 6 months when stored 2-8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seung-Hyun Kim et al.
Oncology letters, 13(6), 4427-4432 (2017-06-11)
Sulforaphane (SFN), an isothiocyanate present in cruciferous vegetables, has been demonstrated to inhibit the growth of various types of cancer cell. The aim of the present study was to investigate whether SFN sensitizes breast cancer cells to paclitaxel-induced apoptosis and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service