Skip to Content
Merck
All Photos(12)

Key Documents

ZRB05665

Sigma-Aldrich

Anti-beta Catenin Antibody, clone 3J7, ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Catenin beta-1

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
R$1,635.00
4 X 25 μL
R$3,368.00

R$1,635.00


In StockDetails


Request a Bulk Order

Select a Size

Change View
25 μL
R$1,635.00
4 X 25 μL
R$3,368.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$1,635.00


In StockDetails


Request a Bulk Order

biological source

rabbit (recombinant)

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

3J7, recombinant monoclonal
monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 85.5 kDa
observed mol wt ~85 kDa

species reactivity

human, mouse

species reactivity (predicted by homology)

rat

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunocytochemistry: suitable using 1:100
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable using 1:100
western blot: suitable using 1:1,000

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... , PRO2286(1499)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 3J7 is a ZooMAb rabbit recombinant monoclonal antibody that detects beta-catenin. It targets an epitope with in the N-terminal region of isoform 1.

Immunogen

GST/His-tagged recombinant fragment corresponding to the first 100 amino acids from the N-terminal region of human beta-catenin.

Application

Anti-beta Catenin, clone 3J7 ZooMAb, Cat. No. ZRB05665, is a recombinant Rabbit Monoclonal Antibody that detects beta-catenin and has been tested for use in Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected beta-Catenin in NIH/3T3 cell lysate.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected beta-Catenin in HUVEC and A431 cells.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected beta- Catenin in human pancreas tissue sections.

Target description

Catenin beta-1 (UniProt: P35222; also known as beta-catenin) is encoded by the CTNNB1 (also known as CTNNB, OK/SW-cl.35, PRO2286) gene (Gene ID: 1499) in human. Catenin beta-1 is a key downstream component of the canonical Wnt signaling pathway that participates in intercellular adhesion and Wnt-mediated transcriptional activation. In the absence of Wnt, it forms a complex with AXIN1, AXIN2, APC, CSNK1A1, and GSK-3 beta that promotes phosphorylation on N-terminal serine and threonine residues that leads to its subsequent degradation by the proteasome. In the presence of Wnt ligand, it is not ubiquitinated and accumulates in the nucleus, where it acts as a coactivator for transcription factors of the TCF/LEF family, which leads to activation of Wnt responsive genes. O-glycosylation at serine 23 is reported to decrease its nuclear localization and transcriptional activity. Wnt/beta-catenin signaling is shown to be essential for embryonic stem cell (ESC) pluripotency and differentiation. In ESC beta-catenin is trimethylated on Lysine 49 by Ezh2 methyltransferase and this trimethylated form represses neuronal differentiation genes Sox1 and Sox3. Mutations in CTNNB1 gene are shown to cause colorectal cancers, medulloblastoma, and ovarian cancers. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Hoffmeyer, K., et al. (2017). Cell Reports 18(12); 2815-2824).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

30 μg/mL after reconstitution at 25 μL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service