Skip to Content
Merck
All Photos(1)

Documents

AB10541

Sigma-Aldrich

Anti-Galectin-3 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Galectin-3, Gal-3, 35 kDa lectin, Carbohydrate-binding protein 35, CBP 35, Galactose-specific lectin 3, Galactoside-binding protein, GALBP, IgE-binding protein, L-31, Laminin-binding protein, Lectin L-29, Mac-2 antigen

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rhesus macaque, human

species reactivity (predicted by homology)

rhesus monkey (based on 100% sequence homology)

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... LGALS3(3958)

General description

Galectin-3 belongs to the galectin gene family, which is defined by the specificity of a carbohydrate recognition domain (CRD) for Gal/Lac. Galectin-3 expression is usually elevated in a wide range of neoplastic cell types. The galectin CRD consists of 135 amino acids. Galectin-3 is known to play a role in tumor development, apoptosis, and cell cycle progression.

Specificity

This antibody recognizes the Pro64 mutation domain of Galectin-3. It is equally specific for wild-type or mutant His64 Galectin-3.

Immunogen

Epitope: Pro64 mutation domain
KLH-conjugated linear peptide corresponding to the Pro64 mutation domain of human Galectin-3.

Application

Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
This Anti-Galectin-3 Antibody is validated for use in WB for the detection of Galectin-3.

Quality

Evaluated by Western Blot in MG-63 cell lysate.

Western Blot Analysis: A 1:250 dilution of this antibody detected Galectin-3 in 10 µg of MG-63 cell lysate.

Target description

~33 kDa observed

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
MG-63 cell lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service