Skip to Content
Merck
All Photos(1)

Key Documents

LM1300

Avanti

12:0-13:0 PS

Avanti Research - A Croda Brand LM1300, methanol solution

Synonym(s):

1-dodecanoyl-2-tridecanoyl-sn-glycero-3-phospho-L-serine (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
R$781.00

R$781.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
R$781.00

About This Item

Empirical Formula (Hill Notation):
C31H63N2O10P
CAS Number:
Molecular Weight:
654.81
UNSPSC Code:
12352211
NACRES:
NA.25

R$781.00


Check Cart for Availability

Request a Bulk Order

form

methanol solution

packaging

pkg of 1 × 1 mL (LM1300-1EA)

manufacturer/tradename

Avanti Research - A Croda Brand LM1300

concentration

~10 μg/mL (Refer to C of A for lot specific concentration.)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OC[C@](C(O)=O)([H])N)=O)(OC(CCCCCCCCCCCC)=O)COC(CCCCCCCCCCC)=O.[NH4+]

InChI

1S/C31H60NO10P.H3N/c1-3-5-7-9-11-13-15-17-19-21-23-30(34)42-27(25-40-43(37,38)41-26-28(32)31(35)36)24-39-29(33)22-20-18-16-14-12-10-8-6-4-2;/h27-28H,3-26,32H2,1-2H3,(H,35,36)(H,37,38);1H3/t27-,28+;/m1./s1

InChI key

DJWXUEZTHOBPTK-CNORLSDQSA-N

General description

Phosphatidylserine is the major anionic phospholipid[1] in human plasma and intracellular membranes identified first in brain.[2] It is a structural component of endoplasmic reticulum, nuclear envelopes, Golgi apparatus, inner layer of plasma membranes and outer mitochondrial membranes. PS is essential for healthy nerve cell membranes and myelin.[2]

Application

12:0-13:0 PS has been used as a quantitative lipid standard to monitor different lipid extracts concentration.[3]

Biochem/physiol Actions

Phosphatidylserine plays a key role in apoptosis by regulating cell signalling molecules and in monitoring membrane fluidity.[1][4] Phosphatidylserine mediates the activation of enzymes and structural components.[1]

Packaging

2 mL Amber Glass Sealed Ampule (LM1300-1EA)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The distribution and function of phosphatidylserine in cellular membranes
Leventis PA and Grinstein S
Annual Review of Biophysics, 39, 407-427 (2010)
The effects of phosphatidylserine on endocrine response to moderate intensity exercise
Starks MA, et al.
Journal of the International Society of Sports Nutrition, 5(1), 11-11 (2008)
A comprehensive method for lipid profiling by liquid chromatography-ion cyclotron resonance mass spectrometry
Fauland A, et al.
Journal of Lipid Research, 52(12), 2314-2322 (2011)
Phosphatidylserine and the human brain
Glade MJ and Smith K
Nutrition, 31(6), 781-786 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service