Skip to Content
Merck
All Photos(1)

Documents

902330

Supelco

Threaded adapter (2") (stainless steel)

with 2 vertical rapid-action connectors

Synonym(s):

Adapter 2" vertical

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121815
NACRES:
NB.51

shipped in

ambient

storage temp.

no temp limit

Related Categories

General description

Our threaded adapters fit in the G2 opening of our stainless steel barrels and drums for solvents. With its 2 different vertical connectors this adapter avoids the mismatch of gas and product side for solvents withdrawal with inert gas pressurizing. The threaded adapter is equipped with Rectus Type 21 male and female as well as with 1 blind plug that can be used with level sensors. (see 1.67710.0001 or 1.67730.0001)
with 2 vertical rapid-action connectors

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service