902335
Dip tube
for use with withdrawal system for stainless steel barrels and drums
Synonym(s):
Dip tube 10 l drum for threaded adapter
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
compatibility
for use with withdrawal system for stainless steel barrels and drums
shipped in
ambient
storage temp.
no temp limit
Related Categories
General description
To customize the withdrawal systems for every requirement, we provide customers with the appropriate dip tubes for each package size suitable for the various withdrawal systems. Dip tube for withdrawal system 1.06710.0001
pour tambour en acier inoxydable de 10 l pour systèmes de prélèvement à adaptateur de filetage 2"
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service