Skip to Content
Merck
All Photos(1)

Key Documents

8.40080

Sigma-Aldrich

Piperonylbutoxide

for synthesis

Synonym(s):

Piperonylbutoxide, α-(2-(2-Butoxyethoxy)ethoxy)-4,5-methylenedioxy-2-propyltoluene, 5-((2-(2-Butoxyethoxy)ethoxy)methyl)-6-propyl-1,3-benzodioxole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H30O5
CAS Number:
Molecular Weight:
338.44
MDL number:
UNSPSC Code:
12352112
EC Index Number:
200-076-7

vapor pressure

0.02 hPa ( 60 °C)

Quality Level

Assay

≥90.0% (GC)

form

liquid

potency

>7000 mg/kg LD50, oral (Rat)
>8000 mg/kg LD50, skin (Rabbit)

bp

180 °C/1.3 hPa

transition temp

flash point 212 °C

solubility

0.01 g/L

density

1.06 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C19H30O5/c1-3-5-7-20-8-9-21-10-11-22-14-17-13-19-18(23-15-24-19)12-16(17)6-4-2/h12-13H,3-11,14-15H2,1-2H3

InChI key

FIPWRIJSWJWJAI-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 90.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.050 - 1.060
Identity (IR): passes test

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

339.8 °F - closed cup

Flash Point(C)

171 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service