Skip to Content
Merck
All Photos(1)

Key Documents

6.17877

Supelco

Perchloric acid

70%, for diamond industry, GR

Synonym(s):

Perchloric acid, PCA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HClO4
CAS Number:
Molecular Weight:
100.46
MDL number:
UNSPSC Code:
12352301

Assay

70%

SMILES string

OCl(=O)(=O)=O

InChI

1S/ClHO4/c2-1(3,4)5/h(H,2,3,4,5)

InChI key

VLTRZXGMWDSKGL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Short-chain polyphosphates: Extraction effects on migration and size estimation of Saccharomyces cerevisiae extracts with polyacrylamide gel electrophoresis.: This study utilized perchloric acid for the extraction of polyphosphates from yeast, which is critical for accurate electrophoresis analysis in biochemical research (Manoukian et al., 2023).
  • Simultaneous determination of 2-(3-hydroxy-5-phosphonooxymethyl-2-methyl-4-pyridyl)-1,3-thiazolidine-4-carboxylic acid and main plasma aminothiols by HPLC-UV based method.: Perchloric acid was employed for sample preparation, crucial for the accurate determination of aminothiols in plasma samples, highlighting its application in clinical chemistry (Piechocka et al., 2023).
  • Determination of Picolinic Acid by HPLC Coupled With Postcolumn Photo Irradiation Using Zinc Acetate as a Fluorescent Derivatization Reagent.: The research utilized perchloric acid in the sample preparation process, demonstrating its importance in biochemical analysis for determining picolinic acid levels (Mawatari et al., 2023).
  • Crystal Structure, Raman, FTIR, UV-Vis Absorption, Photoluminescence Spectroscopy, TG-DSC and Dielectric Properties of New Semiorganic Crystals of 2-Methylbenzimidazolium Perchlorate.: This study employed perchloric acid in the synthesis of new semiorganic crystals, essential for material science research and the characterization of novel compounds (Balashova et al., 2023).
  • Development and validation of a novel HPLC-UV method for simultaneous determination of azathioprine metabolites in human red blood cells.: Perchloric acid was used in the sample preparation for HPLC-UV analysis, crucial for the pharmacokinetic studies of azathioprine metabolites in clinical research (Yu et al., 2023).

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Ox. Liq. 1 - Skin Corr. 1A - STOT RE 2

Target Organs

Thyroid

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service