Perchloric acid
Synonym(s):
PCA, Perchloric acid, Perchloric acid solution
Linear Formula:
HClO4
CAS Number:
Molecular Weight:
100.46
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Purity
Agency
Quality Segment
Reagent Type
Special Grade
Technique
Showing 1-6 of 6
Compare
Product Number
Description
Pricing