Skip to Content
Merck
All Photos(1)

Key Documents

376817-M

Millipore

Hexokinase II Inhibitor II, 3-BP

The Hexokinase II Inhibitor II, 3-BP, also referenced under CAS 1113-59-3, controls the biological activity of Hexokinase II. This small molecule/inhibitor is primarily used for Cell Structure applications.

Synonym(s):

Hexokinase II Inhibitor II, 3-BP, 3-BrPA, Bromopyruvate, Bromopyruvatic Acid, 3-Bromo-2-oxopropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H3BrO3
CAS Number:
Molecular Weight:
166.96
MDL number:
UNSPSC Code:
12352200
EC Index Number:
214-206-5
Pricing and availability is not currently available.

description

RTECS - UZ0840000

Quality Level

Assay

≥97% (titration)

form

crystalline solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

DMSO: 100 mg/mL
distilled water: 100 mg/mL

storage temp.

2-8°C

SMILES string

BrCC(=O)C(=O)O

InChI

1S/C3H3BrO3/c4-1-2(5)3(6)7/h1H2,(H,6,7)

InChI key

PRRZDZJYSJLDBS-UHFFFAOYSA-N

General description

A cell-permeable pyruvic acid derivative that acts as an inhibitor against hexokinase II-mediated glycolysis and effectively suppress tumor survival both in cultures in vitro and in mice and rats in vivo.

Warning

Toxicity: Corrosive (L)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Bujis, M., et al. 2009. Invest. New Drugs27, 120.
Chen, Z., et al. 2009. Biochim. Biophys. Acta.1787, 553.
Hulleman, E., et al. 2009. Blood.113, 2014;
Mathupala, S.P., et al. 2009. Semin. Cancer Biol.19, 17.
Pereira da Silva, A.P., et al. 2009. Biochem. J.417, 717.
Yun, J., et al. 2009. Science.325, 1555.
Zhang, X., et al. 2009. Anticancer Res.29, 1443.
Kim, J.S., et al. 2008. J. Bioenerg. Biomembr.40, 607.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service