Skip to Content
Merck
All Photos(1)

Documents

1.05648

Supelco

HPTLC plates, Silica gel 60 F₂₅₄ Premium Purity

pkg of 50 plates, plate L × W 20 cm × 10 cm, glass support

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711
NACRES:
NB.21

material

glass support
silica gel 60 matrix

Quality Level

feature

binder Organic Polymer
fluorescent indicator

packaging

pkg of 50 plates

technique(s)

thin layer chromatography (TLC): suitable

layer thickness

200 μm

plate L × W

20 cm × 10 cm

particle size

5-6 μm

pore size

60 Å medium pore diameter

storage temp.

2-30°C

General description

50 Glass plates 20 x 10 cm
Ideal for advanced pharmacopoeia applications, this high-performance plate keeps separations free of contamination. The plate is highly pure and exhibits minimal background, even with middle-polar solvent systems. Its separation performance is an exact match to the related HPTLC plate product. Based on the proven HPTLC 60 F254 plate, the HPTLC premium purity plate comes carefully wrapped in a special, plastic-coated aluminum foil. This prevents plasticizers (such as phthalates) from leaving deposits. Without the foil, the plasticizers could appear as an “unknown extra zone” when mid-polar solvent systems (such as toluene/ethyl acetate (95/5)) are in use, and they could be stained by derivatization reagents (such as anis aldehyde).

Application

Glass HPTLC Silica gel 60 F₂₅₄ Premium Purity plates. Silica gel HPTLC plates size 20 x 10 cm, 50 plates.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service