Skip to Content
Merck
All Photos(2)

Documents

1.02071

SAFC

Calcium carbonate

for optical glass

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CaCO3
CAS Number:
Molecular Weight:
100.09
Beilstein:
8008338
MDL number:
UNSPSC Code:
12352302
EC Index Number:
207-439-9

Quality Level

form

solid

potency

6450 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rat)

particle size

75 μm

pH

9.5-10.5 (20 °C, 100 g/L in H2O, slurry)

mp

825 °C (decomposition)

solubility

0.017 g/L

density

2.93 g/mL at 25 °C (lit.)

bulk density

300‑1400 kg/m3

storage temp.

2-30°C

SMILES string

[Ca++].[O-]C([O-])=O

InChI

1S/CH2O3.Ca/c2-1(3)4;/h(H2,2,3,4);/q;+2/p-2

InChI key

VTYYLEPIZMXCLO-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

for optical glass

Application

Calcium carbonate for optical glass. CAS 471-34-1, chemical formula CaCO₃.

Analysis Note

Assay (complexometric)≥ 99.0 %Cr (Chromium)≤ 0.00007 %Fe (Iron)≤ 0.0007 %Particle size (< 75 µm)about 100

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service