1.02069
Calcium carbonate
precipitated, EMPROVE® ESSENTIAL, Ph. Eur., BP, USP, FCC, E 170
Pharma Manufacturing
About This Item
Recommended Products
Agency
BP
Ph. Eur.
USP
Quality Level
reg. compliance
FCC
product line
EMPROVE® ESSENTIAL
form
solid
potency
6450 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rat)
pH
9.5-10.5 (20 °C, 100 g/L in H2O, slurry)
mp
825 °C (decomposition)
density
2.93 g/mL at 25 °C (lit.)
application(s)
liquid formulation
pharmaceutical
solid formulation
storage temp.
2-30°C
SMILES string
[Ca++].[O-]C([O-])=O
InChI
1S/CH2O3.Ca/c2-1(3)4;/h(H2,2,3,4);/q;+2/p-2
InChI key
VTYYLEPIZMXCLO-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
General description
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.
Application
Legal Information
Storage Class Code
13 - Non Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
This article describes the use of excipients to help address API stability issues.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service