Skip to Content
Merck
All Photos(1)

Key Documents

S-058

Supelco

AM-2201 4-Hydroxypentyl metabolite-D5 (indole-D5) solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

AM-2201 Metabolite-D₅ Solution

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
R$979.00

R$979.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
R$979.00

About This Item

Empirical Formula (Hill Notation):
C24H17D5FNO2
Molecular Weight:
380.47
UNSPSC Code:
41116107
NACRES:
NA.24

R$979.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule E (Switzerland)

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology
forensics and toxicology

format

single component solution

storage temp.

−20°C

General description

This internal standard is suited for quantification of AM2201 4-Hydroxypentyl metabolite levels in GC/MS or LC/MS synthetic cannabinoid testing methods for applications in forensic analysis, clinical toxicology, or urine drug testing. AM2201 4-Hydroxypentyl metabolite is a major urinary biomarker of AM2201, a synthetic cannabinoid found in blends of the herbal mixture Spice.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service