Skip to Content
Merck
All Photos(1)

Documents

SMB01373

Sigma-Aldrich

2-Hydroxyesterone

≥95% (HPLC)

Synonym(s):

2-Hydroxyesterone, 2,3-Dihydroxyestra-1,3,5(10)-trien-17-one, 2-Hydroxy estrone, 2-OH-E1, 2-OHE2, 2OHE1, Catecholestrone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H22O3
CAS Number:
Molecular Weight:
286.37
MDL number:
UNSPSC Code:
51351610
NACRES:
NA.21

biological source

synthetic

Quality Level

Assay

≥95% (HPLC)

form

solid

storage temp.

2-8°C

InChI

1S/C18H22O3/c1-18-7-6-11-12(14(18)4-5-17(18)21)3-2-10-8-15(19)16(20)9-13(10)11/h8-9,11-12,14,19-20H,2-7H2,1H3/t11-,12+,14-,18-/m0/s1

InChI key

SWINWPBPEKHUOD-JPVZDGGYSA-N

Application

2-Hydroxyestrone (2OHE1) is a metabolite of the endogenous estrogen estrone. 2OHE1 is formed predominantly by the action of cytochrome P450 (CYP) enzymes CYP1A1, CYP1A2, and CYP1B1. 2OHE1 and 16OHE1 are reported to be associated with breast carcinogenesis.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service