Skip to Content
Merck
All Photos(1)

Key Documents

N-081

Supelco

6β-Naltrexol-D3 solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

6β-Naltrexol-d3

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
R$1,854.00

R$1,854.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
R$1,854.00

About This Item

Empirical Formula (Hill Notation):
C20H22D3NO4
CAS Number:
Molecular Weight:
346.44
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

R$1,854.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

[2H]C([2H])(N1CC[C@]23[C@H]4Oc5c(O)ccc(C[C@@H]1[C@]2(O)CC[C@H]4O)c35)C6([2H])CC6

InChI

1S/C20H25NO4/c22-13-4-3-12-9-15-20(24)6-5-14(23)18-19(20,16(12)17(13)25-18)7-8-21(15)10-11-1-2-11/h3-4,11,14-15,18,22-24H,1-2,5-10H2/t14-,15-,18+,19+,20-/m1/s1/i10D2,11D

InChI key

JLVNEHKORQFVQJ-ZNMQZXPKSA-N

General description

6β-Naltrexol is the primary urinary metabolite of Naltrexone, an opiate sold under the trade names Revia, Depade, and Vivitrol® and used primarily in the management of alcohol or opiate dependence. This internal standard is suitable for 6β-Naltrexol testing or isotope dilution methods in pharmaceutical research, forensic analysis, clinical toxicology, pain prescription monitoring or urine drug testing.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
Vivitrol is a registered trademark of Alkermes, Inc.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service