Skip to Content
Merck
All Photos(1)

Key Documents

IMPN-007-02

Supelco

10-Hydroxynaltrexone solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

10-Hydroxynaltrexone solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H23NO5
CAS Number:
Molecular Weight:
357.40
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

OC1[C@H]2N(CC[C@@]34[C@@H](Oc5c(O)ccc1c35)C(=O)CC[C@@]24O)CC6CC6

InChI

1S/C20H23NO5/c22-12-4-3-11-14-16(12)26-18-13(23)5-6-20(25)17(15(11)24)21(9-10-1-2-10)8-7-19(14,18)20/h3-4,10,15,17-18,22,24-25H,1-2,5-9H2/t15?,17-,18+,19+,20-/m1/s1

InChI key

SPLLFLBLAXVPPQ-KNQKJSHASA-N

General description

10-Hydroxymorphine is a potential impurity from the production of morphine pharmaceutical formulations. Morphine is a potent opiate analgesic drug used to relieve both acute and chronic severe pain. This Certified Snap-N-Spike® Solution is applicable for use as starting material in calibrators or controls for a variety of LC/MS or GC/MS applications such as impurity identification.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service