Skip to Content
Merck
All Photos(1)

Key Documents

LM6004

Avanti

d5-DG ISTD Mix II

Avanti Research - A Croda Brand LM6004

Synonym(s):

d5-DG ISTD Mix II, d5-Diglyceride Mixture Internal Standard, toluene:methanol (1:1)

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
R$3,036.00

R$3,036.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
R$3,036.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.25

R$3,036.00


Check Cart for Availability

Request a Bulk Order

form

solution

packaging

pkg of 1 × 1 mL (LM6004-1EA)

manufacturer/tradename

Avanti Research - A Croda Brand LM6004

concentration

~10 μg/mL (Refer to C of A for lot specific concentration. )

application(s)

lipidomics
metabolomics

shipped in

dry ice

storage temp.

−20°C

General description

Deuterated diglyceride (DG) Mixture II contains: 1,3-16:1 D5 DG 1,3-18:0 D5 DG 1,3-18:1 D5 DG 1,3-18:2 D5 DG.

Application

d5-DG ISTD Mix II or d5-Diglyceride Mixture Internal Standard has been used as an internal standard mixture for diacylglycerol analysis by high-performance liquid chromatography/tandem mass spectrometry (LC/MS2).[1] It has also been used as an internal standard mixture in lipid extraction from 3-hydroxy-isobutyrate (3-HIB)-fed mouse muscle samples and in liquid chromatography-mass spectrometry (LC-MS) lipidomics.[2]

Packaging

2 mL Amber Glass Sealed Ampule (LM6004-1EA)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 1 - STOT SE 3

Target Organs

Central nervous system, Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diminished anabolic signaling response to insulin induced by intramuscular lipid accumulation is associated with inflammation in aging but not obesity
Rivas DA, et al.
American Journal of Physiology. Regulatory, Integrative and Comparative Physiology, 310(7), R561-R569 (2016)
R C Fitzgerald et al.
The Journal of clinical investigation, 98(9), 2120-2128 (1996-11-01)
Barrett's esophagus (BE), or specialized intestinal metaplasia, is a premalignant heterogeneous epithelium associated with reflux and an increased risk for adenocarcinoma. Since acid is a major component of refluxate, we investigated its effects ex vivo on cell differentiation as determined
Cholsoon Jang et al.
Nature medicine, 22(4), 421-426 (2016-03-08)
Epidemiological and experimental data implicate branched-chain amino acids (BCAAs) in the development of insulin resistance, but the mechanisms that underlie this link remain unclear. Insulin resistance in skeletal muscle stems from the excess accumulation of lipid species, a process that
Donato A Rivas et al.
American journal of physiology. Regulatory, integrative and comparative physiology, 310(7), R561-R569 (2016-01-15)
The loss of skeletal muscle mass is observed in many pathophysiological conditions, including aging and obesity. The loss of muscle mass and function with aging is defined as sarcopenia and is characterized by a mismatch between skeletal muscle protein synthesis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service