Skip to Content
Merck
All Photos(1)

Key Documents

330820L

Avanti

UltimateSPLASH ONE

Avanti Research - A Croda Brand

Synonym(s):

UltimateSPLASH ONE Internal Standard for Lipidomic Analysis

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
R$13,581.00

R$13,581.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EACH
R$13,581.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

R$13,581.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

solution

packaging

package of 1 × 1.2 mL

manufacturer/tradename

Avanti Research - A Croda Brand

application(s)

lipidomics

shipped in

dry ice

storage temp.

−20°C

Packaging

2 mL Amber Glass Sealed Ampule (330820L-1EA)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
SPLASH is a trademark of Avanti Polar Lipids, LLC

Kit Components Also Available Separately

Product No.
Description
SDS

  • 860906L14:0-13:0-14:0 TG-d5, Avanti Research - A Croda Brand 25μg/mLSDS

  • 860907L14:0-15:1-14:0 TG-d5, Avanti Research - A Croda Brand 50μg/mLSDS

  • 860908L14:0-17:1-14:0 TG-d5, Avanti Research - A Croda Brand 75μg/mLSDS

  • 860910L16:0-15:1-16:0 TG-d5, Avanti Research - A Croda Brand 100μg/mLSDS

  • 860909L16:0-17:1-16:0 TG-d5, Avanti Research - A Croda Brand 125μg/mLSDS

  • 860911L16:0-19:2-16:0 TG-D5, Avanti Research - A Croda Brand 100μg/mLSDS

  • 860914L18:1-17:1-18:1 TG-d5, Avanti Research - A Croda Brand 75μg/mLSDS

  • 860912L18:1-19:2-18:1 TG-d5, Avanti Research - A Croda Brand 50μg/mLSDS

  • 860913L18:1-21:2-18:1 TG-D5, Avanti Research - A Croda Brand 25μg/mLSDS

  • 700220L14:1 cholesteryl-d7 ester, Avanti Research - A Croda Brand 700220L-1MG 25μg/mLSDS

  • 700221L16:1 cholesteryl-d7 ester, Avanti Research - A Croda Brand 50μg/mLSDS

  • 700222L18:1 cholesteryl-d7 ester, Avanti Research - A Croda Brand 75μg/mLSDS

  • 700223L20:3 Cholestryl-D7 ester, Avanti Research - A Croda Brand 50μg/mLSDS

  • 700226L22:4 Cholesteryl-d7 ester, Avanti Research - A Croda Brand 25μg/mLSDS

  • 860748LC16:1 Ceramide-d7 (d18:1-d7/16:1), Avanti Research - A Croda Brand 75μg/mLSDS

  • 860747LC18:1 Ceramide-d7 (d18:1-d7/18:1), Avanti Research - A Croda Brand 50μg/mLSDS

  • 860746LC20:1 Ceramide-d7 (d18:1-d7/20:1), Avanti Research - A Croda Brand 25μg/mLSDS

  • 860745LC22:1 Ceramide-d7 (d18:1-d7/22:1), Avanti Research - A Croda Brand 50μg/mLSDS

  • 860679LC24:1 Ceramide-d7 (d18:1-d7/24:1), Avanti Research - A Croda Brand 860679L-1MG
    860746L-1MG 75μg/mL
    SDS

  • 860741L16:1 SM (d18:1/16:1)-d9, Avanti Research - A Croda Brand 75μg/mLSDS

  • 860740L18:1 SM (d18:1/18:1)-d9, Avanti Research - A Croda Brand 860740L-1MG
    860746L-1MG 50μg/mL
    SDS

  • 860742L20:1 SM (d18:1/20:1)-d9, Avanti Research - A Croda Brand 25μg/mLSDS

  • 860743L22:1 SM (d18:1/22:1)-d9, Avanti Research - A Croda Brand 50μg/mLSDS

  • 860744L24:1 SM (d18:1/24:1)-d9, Avanti Research - A Croda Brand 75μg/mLSDS

  • 855683L17:0-14:1 PC-d5, Avanti Research - A Croda Brand 50μg/mLSDS

  • 855682L17:0-16:1 PC-d5, Avanti Research - A Croda Brand 100μg/mLSDS

  • 855681L17:0-18:1 PC-d5, Avanti Research - A Croda Brand 150μg/mLSDS

  • 855680L17:0-20:3 PC-d5, Avanti Research - A Croda Brand 100μg/mLSDS

  • 855678L17:0-22:4 PC-d5, Avanti Research - A Croda Brand 50μg/mLSDS

  • 856721L17:0-14:1 PE-d5, Avanti Research - A Croda Brand 25μg/mLSDS

  • 856720L17:0-16:1 PE-d5, Avanti Research - A Croda Brand 50μg/mLSDS

  • 856719L17:0-18:1 PE-d5, Avanti Research - A Croda Brand 75μg/mLSDS

  • 856718L17:0-20:3 PE-d5, Avanti Research - A Croda Brand 50μg/mLSDS

  • 856717L17:0-22:4 PE-d5, Avanti Research - A Croda Brand 25μg/mLSDS

  • 858135L17:0-14:1 PG-d5, Avanti Research - A Croda Brand 858135L-1MG
    860746L-1MG 25μg/mL
    SDS

  • 858134L17:0-16:1 PG-d5, Avanti Research - A Croda Brand 50μg/mLSDS

  • 858133L17:0-18:1 PG-d5, Avanti Research - A Croda Brand 75μg/mLSDS

  • 858132L17:0-20:3 PG-d5, Avanti Research - A Croda Brand 50μg/mLSDS

  • 858131L17:0-22:4 PG-d5, Avanti Research - A Croda Brand 25μg/mLSDS

  • 858153L17:0-14:1 PS-d5, Avanti Research - A Croda Brand 25μg/mLSDS

  • 858152L17:0-16:1 PS-d5, Avanti Research - A Croda Brand 50μg/mLSDS

  • 858151L17:0-18:1 PS-d5, Avanti Research - A Croda Brand 75μg/mLSDS

  • 858150L17:0-20:3 PS-d5, Avanti Research - A Croda Brand 50μg/mLSDS

  • 858149L17:0-22:4 PS-d5, Avanti Research - A Croda Brand 25μg/mLSDS

  • 800827L17:0-14:1 DG-d5, Avanti Research - A Croda Brand 25μg/mLSDS

  • 800826L17:0-16:1 DG-d5, Avanti Research - A Croda Brand 50μg/mLSDS

  • 800824L17:0-18:1 DG-d5, Avanti Research - A Croda Brand 75μg/mLSDS

  • 800825L17:0-20:3 DG-d5, Avanti Research - A Croda Brand 50μg/mLSDS

  • 800823L17:0-22:4 DG-d5, Avanti Research - A Croda Brand 25μg/mLSDS

  • 850109L17:0-14:1 PI-d5, Avanti Research - A Croda Brand 25μg/mLSDS

  • 850110L17:0-16:1 PI-d5, Avanti Research - A Croda Brand 50μg/mLSDS

  • 850111L17:0-18:1 PI-d5, Avanti Research - A Croda Brand 850111L-500UG 75μg/mLSDS

  • 850112L17:0-20:3 PI-d5, Avanti Research - A Croda Brand 50μg/mLSDS

  • 850118L17:0-22:4 PI-d5, Avanti Research - A Croda Brand 25μg/mLSDS

  • 850107L15:0 Lyso PI-d5, Avanti Research - A Croda Brand 25μg/mLSDS

  • 850108L17:0 Lyso PI-d5, Avanti Research - A Croda Brand 50μg/mLSDS

  • 850106L19:0 LYSO PI-d5, Avanti Research - A Croda Brand 850106L-500UG 25μg/mLSDS

  • 858146L15:0 Lyso PS-d5, Avanti Research - A Croda Brand 858146L-1MG 25μg/mLSDS

  • 858148L17:0 Lyso PS-d5, Avanti Research - A Croda Brand 50μg/mLSDS

  • 858147L19:0 Lyso PS-d5, Avanti Research - A Croda Brand 25μg/mLSDS

  • 858123L15:0 Lyso PG-d5, Avanti Research - A Croda Brand 25μg/mLSDS

  • 858130L17:0 Lyso PG-d5, Avanti Research - A Croda Brand 50μg/mLSDS

  • 858129L19:0 LYSO PG-d5, Avanti Research - A Croda Brand 25μg/mLSDS

  • 870309L15:0 Lyso PC-d5, Avanti Research - A Croda Brand 25μg/mLSDS

  • 855679L17:0 Lyso PC-d5, Avanti Research - A Croda Brand 50μg/mLSDS

  • 855778L19:0 Lyso PC-d5, Avanti Research - A Croda Brand 855778L-1MG 25μg/mLSDS

  • 856709L15:0 Lyso PE-d5, Avanti Research - A Croda Brand 25μg/mLSDS

  • 856710L17:0 Lyso PE-d5, Avanti Research - A Croda Brand 50μg/mLSDS

  • 856716L19:0 Lyso PE-d5, Avanti Research - A Croda Brand 25μg/mLSDS

See All (69)

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 1 - STOT SE 3

Target Organs

Central nervous system, Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service