Skip to Content
Merck
All Photos(1)

Key Documents

880127C

Avanti

DSPE-PEG(2000) PDP

Avanti Research - A Croda Brand 880127C

Synonym(s):

1,2-distearoyl-sn-glycero-3-phosphoethanolamine-N-[PDP(polyethylene glycol)-2000] (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
R$3,724.00

R$3,724.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
R$3,724.00

About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

R$3,724.00


Check Cart for Availability

Request a Bulk Order

form

liquid

packaging

pkg of 1 × 2.5 mL (880127C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 880127C

concentration

10 mg/mL (880127C-25mg)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCNC(OCCOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOO/C=C\NC(CCSSC1=CC=CC=N1)=O)=O)=O)(OC(CCCCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCCCC)=O.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.

General description

DSPE-PEG(2000) PDP is a functionalized polyethylene glycol (PEG) conjugated lipid.

Application

DSPE-PEG(2000) PDP has been used in the preparation of targeted liposomes.[1] It has also been used as a PEGylated lipid for antibody conjugation to vesicles.[2]

Biochem/physiol Actions

DSPE-PEG(2000) PDP serves as a precursor of DSPE-PEG2000-SH.[3]

Packaging

5 mL Clear Glass Sealed Ampule (880127C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The use of pH-triggered leaky heterogeneities on rigid lipid bilayers to improve intracellular trafficking and therapeutic potential of targeted liposomal immunochemotherapy
Karve S, et al.
Biomaterials, 30(30), 6055-6064 (2009)
Antitumor efficacy following the intracellular and interstitial release of liposomal doxorubicin
Bandekar A, et al.
Biomaterials, 33(17), 4345-4352 (2012)
Multifunctional nanoliposomes with curcumin--lipid derivative and brain targeting functionality with potential applications for Alzheimer disease
Mourtas S, et al.
European Journal of Medicinal Chemistry, 80, 175-183 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service