Skip to Content
Merck
All Photos(1)

Key Documents

880127P

Avanti

DSPE-PEG(2000) PDP

Avanti Research - A Croda Brand 880127P, powder

Synonym(s):

1,2-distearoyl-sn-glycero-3-phosphoethanolamine-N-[PDP(polyethylene glycol)-2000] (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
R$875.00
25 MG
R$1,873.00
100 MG
R$5,887.00

R$875.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
R$875.00
25 MG
R$1,873.00
100 MG
R$5,887.00

About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

R$875.00


Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 10 mg (880127P-10mg)
pkg of 1 × 100 mg (880127P-100mg)
pkg of 1 × 25 mg (880127P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 880127P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCNC(OCCOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOO/C=C\NC(CCSSC1=CC=CC=N1)=O)=O)=O)(OC(CCCCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCCCC)=O.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.

General description

DSPE-PEG(2000) PDP is a functionalized polyethylene glycol (PEG) conjugated lipid.

Application

DSPE-PEG(2000) PDP has been used as a standard to study the structural and functional organization of synthesized tethered bilayer lipid membranes.[1]

Biochem/physiol Actions

DSPE-PEG(2000) PDP serves as a precursor of DSPE-PEG2000-SH.[2]

Packaging

5 mL Amber Glass Screw Cap Vial (880127P-100mg)
5 mL Amber Glass Screw Cap Vial (880127P-10mg)
5 mL Amber Glass Screw Cap Vial (880127P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solid supported lipid membranes: New concepts for the biomimetic functionalization of solid surfaces
Knoll W, et al.
Biointerphases, 3(2), FA125-FA135 (2008)
Multifunctional nanoliposomes with curcumin--lipid derivative and brain targeting functionality with potential applications for Alzheimer disease
Mourtas S, et al.
European Journal of Medicinal Chemistry, 80, 175-183 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service