Skip to Content
Merck
All Photos(1)

Documents

870129C

Avanti

18:1 DBCO PE

Avanti Research - A Croda Brand 870129C

Synonym(s):

18:1 DBCO PE, 1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-dibenzocyclooctyl

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

liquid

packaging

pkg of 1 × 2.5 mL (870129C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870129C

concentration

10 mg/mL (870129C-25mg)

lipid type

headgroup modified lipids
click reagents

shipped in

dry ice

storage temp.

−20°C

General description

18:1 DBCO PE, also known as 1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-dibenzocyclooctyl, is a head group modified lipid containing dibenzocyclooctyne (DBCO). It is mostly used as a click reagent.

Packaging

5 mL Clear Glass Sealed Ampule (870129C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service