Skip to Content
Merck
All Photos(1)

Key Documents

870128P

Avanti

16:0 DBCO PE

Avanti Research - A Croda Brand 870128P, powder

Synonym(s):

1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine-N-dibenzocyclooctyl

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
R$1,873.00

R$1,873.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
R$1,873.00

About This Item

Empirical Formula (Hill Notation):
C56H90N3O10P
CAS Number:
Molecular Weight:
996.30
UNSPSC Code:
12352211
NACRES:
NA.25

R$1,873.00


Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 25 mg (870128P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870128P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCNC(CCC(N1CC(C=CC=C2)=C2C#CC3=C1C=CC=C3)=O)=O)=O)(OC(CCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O.[NH4+]

InChI key

VMNQBMCGXSXSJG-GLOFXQGCSA-N

General description

1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine-N-dibenzocyclooctyl (16:0 DBCO PE) is a head group modified lipid containing dibenzocyclooctyne (DBCO). It is a widely used reagent in click chemistry.[1]

Application

1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine-N-dibenzocyclooctyl (16:0 DBCO PE or DPPE-DBCO) has been used in preparation of DBCO-presenting lipid bilayers.[2] It has also been used in preparation of clickable cubosomes for antibody-free drug targeting and imaging applications.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (870128P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicolas Alcaraz et al.
Bioconjugate chemistry, 29(1), 149-157 (2017-11-29)
The combination of copper-free click chemistry with metabolic labeling offers new opportunities in drug delivery. The objective of this study was to determine whether cubosomes functionalized with azide or dibenzocyclooctyne (DBCO) groups are able to undergo copper-free click chemistry with
Simon Finn Mayer et al.
Biochimica et biophysica acta. Biomembranes, 1861(10), 183023-183023 (2019-07-22)
Pore-forming antimicrobial peptides (AMPs) are attracting interest as cytolytic antibiotics and drug delivery agents with potential use for targeting cancer cells or multidrug-resistant pathogens. Ceratotoxin A (CtxA) is an insect-derived cytolytic AMP with 36 amino acids that is thought to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service