Skip to Content
Merck
All Photos(1)

Key Documents

791648C

Avanti

15:0-18:1-d7-15:0 TG

Avanti Research - A Croda Brand 791648C

Synonym(s):

1,3-dipentadecanoyl-2-oleyol(d7)-glycerol

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
R$1,094.00

R$1,094.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
R$1,094.00

About This Item

Empirical Formula (Hill Notation):
C51H89 D7O6
CAS Number:
Molecular Weight:
812.35
MDL number:
UNSPSC Code:
41141833
NACRES:
NA.25

R$1,094.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (791648C-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 791648C

concentration

1 mg/mL (791648C-1mg)

shipped in

dry ice

storage temp.

−20°C

General description

This deuterium labeled lipid is also available in a formulation of multiple lipid standards. The SPLASH LIPIDOMIX Standard includes all of the major lipid classes at ratios relative to human plasma, allowing one easy internal standard solution to be added to every sample.

Application

15:0-18:1-d7-15:0 TG may be used as an internal standard in the high performance liquid chromatography-electrospray ionization triple quadrupole mass spectrometry (HPLC-ESI-QqQ-MS).[1]

Packaging

2 mL Amber Glass Sealed Ampule (791648C-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
LIPIDOMIX is a trademark of Avanti Polar Lipids, LLC
SPLASH is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Food targeting: Geographical origin determination of hazelnuts (Corylus avellana) by LC-QqQ-MS/MS-based targeted metabolomics application
Klockmann S, et al.
Journal of Agricultural and Food Chemistry, 65(7), 1456-1465 (2017)
Ya Xie et al.
Food chemistry, 319, 126547-126547 (2020-03-18)
In this research, a novel ultrasound-assisted extraction (USAE) method using a one-phase solvent (isopropanol, IPA) combined with liquid chromatography-electrospray ionization quadrupole time-of-flight mass spectrometry (LC-ESI-QToF-MS) was developed for exhaustive profiling of egg yolk lipids, which are considered to be essential

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service