Skip to Content
Merck
All Photos(1)

Key Documents

791643C

Avanti

18:1-d7 Lyso PC

Avanti Research - A Croda Brand

Synonym(s):

1-oleoyl(d7)-2-hydroxy-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
R$1,094.00

R$1,094.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
R$1,094.00

About This Item

Empirical Formula (Hill Notation):
C26H45D7NO7P
CAS Number:
Molecular Weight:
528.71
MDL number:
UNSPSC Code:
41141825
NACRES:
NA.25

R$1,094.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (791643C-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

1 mg/mL (791643C-1mg)

shipped in

dry ice

storage temp.

−20°C

General description

18:1-d7 Lyso PC 1-oleoyl(d7)-2-hydroxy-sn-glycero-3-phosphocholine is a deuterated phospholipid. The lipid lysophosphatidylcholine is esterified with oleic acid. Oleic acid harbors the deuterium ions. Lysophosphatidylcholine (LPC) is derived from phosphatidylcholine by the action of enzyme phospholipase A2.[1]
This deuterium labeled lipid is also available in a formulation of multiple lipid standards. The SPLASH LIPIDOMIX Standard includes all of the major lipid classes at ratios relative to human plasma, allowing one easy internal standard solution to be added to every sample.

Application

18:1-d7 Lyso PC has been used:
  • as an internal standard in high performance liquid chromatography electrospray ionization tandem mass spectrometry (HPLC-ESIMS/MS) for the lipidomics analysis of bone marrow-derived macrophages[2]
  • as a component of extraction solvent for lipid isolation from biological samples[3]
  • as an internal standard for spiking serum samples for liquid chromatography with tandem mass spectrometry (LC-MS/MS)[4]

Biochem/physiol Actions

Lysophosphatidylcholine (LPC). Elevated levels of LPC is associated with diabetes and cardiovascular diseases.[1] LPC stimulates cytokines and is implicated in allergic disease. It also elicits lysis on membranes.[5]

Packaging

5 mL Clear Glass Sealed Ampule (791643C-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
LIPIDOMIX is a trademark of Avanti Polar Lipids, LLC
SPLASH is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preeti Bansal et al.
Scientific reports, 6, 27430-27430 (2016-06-11)
Phospholipase A2 (sPLA2), pivotal for allergic and inflammatory response, hydrolyses phosphatidylcholine (PC) to lysophosphatidylcholine (LPC). In present study, the role of LPC in allergic airway disease manifestation was studied using mouse model. Balb/c mice were immunized using cockroach extract (CE)
Shi-Hui Law et al.
International journal of molecular sciences, 20(5) (2019-03-09)
Lysophosphatidylcholine (LPC) is increasingly recognized as a key marker/factor positively associated with cardiovascular and neurodegenerative diseases. However, findings from recent clinical lipidomic studies of LPC have been controversial. A key issue is the complexity of the enzymatic cascade involved in
Shayne A Snider et al.
The Journal of biological chemistry, 293(29), 11600-11611 (2018-06-09)
Choline is an essential nutrient that is required for synthesis of the main eukaryote phospholipid, phosphatidylcholine. Macrophages are innate immune cells that survey and respond to danger and damage signals. Although it is well-known that energy metabolism can dictate macrophage
Michael S Gardner et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1118-1119, 137-147 (2019-04-30)
Progress toward better diagnosis and treatment of lipid metabolism-related diseases requires high throughput approaches for multiplexed quantitative analysis of structurally diverse lipids, including phospholipids (PLs). This work demonstrates a simplified "one-pot" phospholipid extraction protocol, as an alternative to conventional liquid-liquid
Tong Liu et al.
Expert review of proteomics, 17(3), 233-242 (2020-04-21)
Background: Colorectal cancer (CRC) represents a third leading cause of cancer-related death worldwide. The reliable diagnostic biomarkers for detecting CRC at early stage is critical for decreasing the mortality.Method: A conjunctive lipidomic approach was employed to investigate the differences in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service