Skip to Content
Merck
All Photos(2)

Documents

700076P

Avanti

8(14)-dehydrocholesterol

Avanti Research - A Croda Brand

Synonym(s):

cholesta-5,8(14)-dien-3β-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H44O
CAS Number:
Molecular Weight:
384.64
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (700076P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−70°C

SMILES string

C[C@@]12C(CCC2C(CCCC(C)C)C)=C3CC=C4C[C@@H](O)CC[C@]4(C)C3CC1

Biochem/physiol Actions

8(14)-dehydrocholesterol is a cholestadienol. The free radical oxidation leads to abstraction of hydrogen at the bis-allylic positions leading to formation of D7-3,5-dihydroxycholest-7-en-6-one (DHCEO).

Packaging

5 mL Amber Glass Screw Cap Vial (700076P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reactivities and products of free radical oxidation of cholestadienols
Xu L and Porter NA
Journal of the American Chemical Society, 136(14), 5443-5450 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service